Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFRSF10C
Entrez Gene ID
8794
Gene Name
TNF receptor superfamily member 10c
Gene Aliases
CD263, DCR1, DCR1-TNFR, LIT, TRAIL-R3, TRAILR3, TRID
Chromosome Location
Chr.8: 23102814 - 23117437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003841.3 NP_003832.2 1 - 2 367 64
GenBank mRNA AF012536.1 - 1 - 2 251 64
AF012629.1 - 1 - 2 59 64
AF014794.1 - 1 - 2 208 64
AF016267.1 - 1 - 2 246 64
AF020502.1 - 1 - 2 179 64
AF033854.1 - 1 - 2 235 64
AK298718.1 - 1 - 2 258 64
AK313213.1 - 1 - 2 261 64
AY358281.1 - 1 - 2 251 64
BC017852.1 - 1 - 2 260 64
BC125041.1 - 1 - 2 180 64
BC125042.1 - 1 - 2 180 64
BG545631.1 - 1 - 2 257 64
CR541857.1 - 1 - 2 59 64
DC380668.1 - 1 - 2 367 64

View Full Product Details