The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014967.4 | NP_055782.3 | 11 - 12 | 2887 | 66 |
XM_005254232.4 | XP_005254289.1 | 11 - 12 | 2895 | 66 | |
XM_005254234.4 | XP_005254291.1 | 11 - 12 | 2941 | 66 | |
XM_005254235.3 | XP_005254292.1 | 11 - 12 | 2777 | 66 | |
XM_011521370.2 | XP_011519672.1 | 11 - 12 | 1580 | 66 | |
XM_011521371.1 | XP_011519673.1 | 12 - 13 | 3006 | 66 | |
XM_017022012.1 | XP_016877501.1 | 10 - 11 | 2657 | 66 | |
XM_017022013.1 | XP_016877502.1 | 10 - 11 | 2670 | 66 | |
GenBank mRNA | AB023235.2 | - | 11 - 12 | 2834 | 66 |
CR936727.1 | - | 12 - 13 | 3001 | 66 | |
CR936734.1 | - | 11 - 12 | 2857 | 66 | |
HM560957.1 | - | 10 - 11 | 2596 | 66 |