The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004053.3 | NP_004044.3 | 1 - 2 | 607 | 111 |
GenBank mRNA | AK314147.1 | - | 1 - 2 | 362 | 111 |
BC007340.2 | - | 1 - 2 | 343 | 111 | |
BC050645.1 | - | 1 - 2 | 607 | 111 | |
BC062627.1 | - | 1 - 2 | 578 | 111 |