The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004053.3 | NP_004044.3 | 3 - 4 | 906 | 95 |
GenBank mRNA | AK095253.1 | - | - | 1326 | 95 |
AK314147.1 | - | 3 - 4 | 661 | 95 | |
BC007340.2 | - | 3 - 4 | 642 | 95 | |
BC050645.1 | - | 3 - 4 | 906 | 95 | |
BC062627.1 | - | 3 - 4 | 877 | 95 | |
L36720.2 | - | 2 - 3 | 186 | 95 |