Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA1211L
Entrez Gene ID
343990
Gene Name
KIAA1211 like
Gene Aliases
C2orf55
Chromosome Location
Chr.2: 98793846 - 98936221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207362.2 NP_997245.2 7 - 8 2710 73
XM_005263923.3 XP_005263980.1 7 - 8 2690 73
XM_005263924.3 XP_005263981.1 7 - 8 2690 73
XM_005263926.3 XP_005263983.1 7 - 8 2575 73
XM_011511093.1 XP_011509395.1 9 - 10 2849 73
XM_011511094.1 XP_011509396.1 8 - 9 2747 73
XM_011511095.2 XP_011509397.1 7 - 8 2517 73
XM_011511096.1 XP_011509398.1 4 - 5 2127 73
XM_017003971.1 XP_016859460.1 7 - 8 2693 73
XM_017003972.1 XP_016859461.1 8 - 9 2797 73

View Full Product Details