Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA1211L
Entrez Gene ID
343990
Gene Name
KIAA1211 like
Gene Aliases
C2orf55
Chromosome Location
Chr.2: 98793846 - 98936221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207362.2 NP_997245.2 3 - 4 531 100
XM_005263923.3 XP_005263980.1 3 - 4 511 100
XM_005263924.3 XP_005263981.1 3 - 4 511 100
XM_005263926.3 XP_005263983.1 3 - 4 396 100
XM_011511093.1 XP_011509395.1 5 - 6 670 100
XM_011511094.1 XP_011509396.1 4 - 5 568 100
XM_011511095.2 XP_011509397.1 3 - 4 338 100
XM_017003971.1 XP_016859460.1 3 - 4 514 100
XM_017003972.1 XP_016859461.1 4 - 5 618 100
XM_017003973.1 XP_016859462.1 3 - 4 524 100
GenBank mRNA BM922097.1 - 3 - 4 549 100

View Full Product Details