Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFXL1
Entrez Gene ID
152518
Gene Name
nuclear transcription factor, X-box binding like 1
Gene Aliases
CDZFP, HOZFP, OZFP, URCC5
Chromosome Location
Chr.4: 47847233 - 47914667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278623.1 NP_001265552.1 18 - 19 2302 117
NM_001278624.1 NP_001265553.1 18 - 19 2440 117
NM_152995.5 NP_694540.3 18 - 19 2342 117
NR_103795.1 19 - 20 2482 117
GenBank mRNA AB085695.1 - 18 - 19 2298 117
AB181916.1 - 18 - 19 2422 117
AK131285.1 - 16 - 17 1888 117
AK302440.1 - 18 - 19 2300 117
AY134856.1 - 2 - 3 346 117
BC051193.1 - 19 - 20 2453 117

View Full Product Details