Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFXL1
Entrez Gene ID
152518
Gene Name
nuclear transcription factor, X-box binding like 1
Gene Aliases
CDZFP, HOZFP, OZFP, URCC5
Chromosome Location
Chr.4: 47847233 - 47914667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278623.1 NP_001265552.1 20 - 21 2484 120
NM_001278624.1 NP_001265553.1 20 - 21 2622 120
NM_152995.5 NP_694540.3 20 - 21 2524 120
NR_103795.1 21 - 22 2664 120
GenBank mRNA AB085695.1 - 20 - 21 2480 120
AB181916.1 - 20 - 21 2604 120
AK131285.1 - 18 - 19 2070 120
AK302440.1 - 20 - 21 2482 120
AY134856.1 - 4 - 5 528 120
BC018019.1 - 3 - 4 219 120
BC051193.1 - 21 - 22 2635 120

View Full Product Details