The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016323.3 | NP_057407.2 | 15 - 16 | 2111 | 122 |
GenBank mRNA | AB027289.1 | - | 15 - 16 | 2071 | 122 |
AK093155.1 | - | 5 - 6 | 1446 | 122 | |
AK301756.1 | - | 5 - 6 | 679 | 122 | |
AK302142.1 | - | 9 - 10 | 983 | 122 | |
AY337518.1 | - | 15 - 16 | 2111 | 122 | |
BC140716.1 | - | 15 - 16 | 2004 | 122 |