Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HERC5
Entrez Gene ID
51191
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 5
Gene Aliases
CEB1, CEBP1
Chromosome Location
Chr.4: 88456604 - 88506170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016323.3 NP_057407.2 7 - 8 1213 86
XM_011532022.2 XP_011530324.2 7 - 8 1726 86
GenBank mRNA AB027289.1 - 7 - 8 1173 86
AY337518.1 - 7 - 8 1213 86
BC140716.1 - 7 - 8 1106 86

View Full Product Details