The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138557.2 | NP_612567.1 | 1 - 2 | 389 | 67 |
GenBank mRNA | AK303730.1 | - | 1 - 2 | 220 | 67 |
BC025294.1 | - | 1 - 2 | 175 | 67 | |
DA187988.1 | - | 1 - 2 | 389 | 67 |