The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004774.3 | NP_004765.2 | 1 - 2 | 237 | 92 |
XM_005257465.3 | XP_005257522.1 | 1 - 2 | 244 | 92 | |
XM_006721957.1 | XP_006722020.1 | 1 - 2 | 234 | 92 | |
XM_017024779.1 | XP_016880268.1 | 1 - 2 | 244 | 92 | |
GenBank mRNA | AF055994.1 | - | 1 - 2 | 217 | 92 |
AK299789.1 | - | 1 - 2 | 235 | 92 | |
AK314185.1 | - | 1 - 2 | 234 | 92 | |
BC006517.2 | - | 1 - 2 | 210 | 92 | |
BC060758.1 | - | 1 - 2 | 215 | 92 | |
BC131783.1 | - | 1 - 2 | 91 | 92 | |
BP357374.1 | - | 1 - 2 | 237 | 92 | |
Y13467.1 | - | 1 - 2 | 215 | 93 |