Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAU2
Entrez Gene ID
23383
Gene Name
MAU2 sister chromatid cohesion factor
Gene Aliases
KIAA0892, MAU2L, SCC4, mau-2
Chromosome Location
Chr.19: 19320686 - 19358754 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015329.3 NP_056144.3 15 - 16 1477 67
XM_005259837.3 XP_005259894.2 15 - 16 1466 67
XM_005259838.3 XP_005259895.3 13 - 14 1528 67
XM_006722711.3 XP_006722774.2 15 - 16 1473 67
XM_011527844.2 XP_011526146.1 15 - 16 1882 67
XM_017026539.1 XP_016882028.1 13 - 14 1524 67
GenBank mRNA AB020699.1 - 15 - 16 1463 67
AK001349.1 - 3 - 4 194 67
AK025896.1 - 13 - 14 1129 67
AK126227.1 - 2 - 3 1369 67
AK128029.1 - 4 - 5 1335 67
AL110250.1 - 4 - 5 252 67
BC010222.1 - 4 - 5 263 67
BC063863.1 - 4 - 5 610 67

View Full Product Details