Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKP4
Entrez Gene ID
8502
Gene Name
plakophilin 4
Gene Aliases
p0071
Chromosome Location
Chr.2: 158456880 - 158681429 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005476.2 NP_001005476.1 16 - 17 3068 127
NM_001304969.1 NP_001291898.1 16 - 17 3065 127
NM_001304970.1 NP_001291899.1 16 - 17 2988 127
NM_003628.4 NP_003619.2 16 - 17 3068 127
XM_011512016.1 XP_011510318.1 16 - 17 3505 127
XM_011512017.1 XP_011510319.1 16 - 17 2809 127
XM_011512018.1 XP_011510320.1 16 - 17 3499 127
XM_011512019.1 XP_011510321.1 16 - 17 2806 127
XM_011512020.2 XP_011510322.1 16 - 17 2852 127
XM_011512021.2 XP_011510323.1 16 - 17 2852 127
XM_011512022.1 XP_011510324.1 15 - 16 3020 127
XM_011512025.1 XP_011510327.1 11 - 12 1940 127
XM_017005122.1 XP_016860611.1 16 - 17 3502 127
XM_017005123.1 XP_016860612.1 16 - 17 4037 127
XM_017005124.1 XP_016860613.1 16 - 17 3496 127
XM_017005125.1 XP_016860614.1 16 - 17 2849 127
XM_017005126.1 XP_016860615.1 16 - 17 2809 127
XM_017005127.1 XP_016860616.1 16 - 17 3505 127
XM_017005128.1 XP_016860617.1 16 - 17 2849 127
XM_017005129.1 XP_016860618.1 15 - 16 3014 127
XM_017005130.1 XP_016860619.1 15 - 16 3017 127
XM_017005131.1 XP_016860620.1 11 - 12 1937 127
XM_017005132.1 XP_016860621.1 14 - 15 2345 127
XM_017005133.1 XP_016860622.1 15 - 16 2472 127
XM_017005134.1 XP_016860623.1 14 - 15 2839 127
XM_017005135.1 XP_016860624.1 13 - 14 2042 127
GenBank mRNA AK124237.1 - 3 - 4 1168 127
AK128720.1 - 14 - 15 2895 127
BC048013.1 - 16 - 17 2902 127
BC050308.1 - 16 - 17 2850 127
X81889.1 - 16 - 17 2865 127

View Full Product Details