Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKP4
Entrez Gene ID
8502
Gene Name
plakophilin 4
Gene Aliases
p0071
Chromosome Location
Chr.2: 158456880 - 158681429 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304969.1 NP_001291898.1 19 - 20 3471 72
NM_001304970.1 NP_001291899.1 19 - 20 3394 72
NM_003628.4 NP_003619.2 19 - 20 3474 72
XM_011512016.1 XP_011510318.1 19 - 20 3911 72
XM_011512017.1 XP_011510319.1 19 - 20 3215 72
XM_011512018.1 XP_011510320.1 19 - 20 3905 72
XM_011512019.1 XP_011510321.1 19 - 20 3212 72
XM_011512020.2 XP_011510322.1 19 - 20 3258 72
XM_011512022.1 XP_011510324.1 18 - 19 3426 72
XM_011512025.1 XP_011510327.1 14 - 15 2346 72
XM_017005122.1 XP_016860611.1 19 - 20 3908 72
XM_017005123.1 XP_016860612.1 19 - 20 4443 72
XM_017005124.1 XP_016860613.1 19 - 20 3902 72
XM_017005129.1 XP_016860618.1 18 - 19 3420 72
XM_017005130.1 XP_016860619.1 18 - 19 3423 72
XM_017005131.1 XP_016860620.1 14 - 15 2343 72
XM_017005132.1 XP_016860621.1 17 - 18 2751 72
XM_017005133.1 XP_016860622.1 18 - 19 2878 72
XM_017005134.1 XP_016860623.1 17 - 18 3245 72
GenBank mRNA AK055015.1 - 3 - 4 952 72
AK124237.1 - 6 - 7 1574 72
BC048013.1 - 19 - 20 3308 72
X81889.1 - 19 - 20 3271 72

View Full Product Details