Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKP4
Entrez Gene ID
8502
Gene Name
plakophilin 4
Gene Aliases
p0071
Chromosome Location
Chr.2: 158456880 - 158681429 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005476.2 NP_001005476.1 1 - 2 346 65
NM_001304969.1 NP_001291898.1 1 - 2 346 65
NM_001304970.1 NP_001291899.1 1 - 2 346 65
NM_001304971.1 NP_001291900.1 1 - 2 346 65
NM_003628.4 NP_003619.2 1 - 2 346 65
XM_011512017.1 XP_011510319.1 1 - 2 93 65
XM_011512019.1 XP_011510321.1 1 - 2 93 65
XM_011512020.2 XP_011510322.1 1 - 2 130 65
XM_011512021.2 XP_011510323.1 1 - 2 130 65
XM_011512026.2 XP_011510328.1 1 - 2 130 65
XM_017005125.1 XP_016860614.1 1 - 2 130 65
XM_017005126.1 XP_016860615.1 1 - 2 93 65
XM_017005128.1 XP_016860617.1 1 - 2 130 65
GenBank mRNA BC050308.1 - 1 - 2 128 65

View Full Product Details