Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKP4
Entrez Gene ID
8502
Gene Name
plakophilin 4
Gene Aliases
p0071
Chromosome Location
Chr.2: 158456880 - 158681429 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005476.2 NP_001005476.1 4 - 5 619 84
NM_001304969.1 NP_001291898.1 4 - 5 619 84
NM_001304970.1 NP_001291899.1 4 - 5 619 84
NM_001304971.1 NP_001291900.1 4 - 5 619 84
NM_003628.4 NP_003619.2 4 - 5 619 84
XM_011512016.1 XP_011510318.1 4 - 5 1056 84
XM_011512017.1 XP_011510319.1 4 - 5 366 84
XM_011512018.1 XP_011510320.1 4 - 5 1056 84
XM_011512019.1 XP_011510321.1 4 - 5 366 84
XM_011512020.2 XP_011510322.1 4 - 5 403 84
XM_011512021.2 XP_011510323.1 4 - 5 403 84
XM_011512022.1 XP_011510324.1 3 - 4 571 84
XM_011512026.2 XP_011510328.1 4 - 5 403 84
XM_017005122.1 XP_016860611.1 4 - 5 1056 84
XM_017005123.1 XP_016860612.1 4 - 5 1591 84
XM_017005124.1 XP_016860613.1 4 - 5 1056 84
XM_017005125.1 XP_016860614.1 4 - 5 403 84
XM_017005126.1 XP_016860615.1 4 - 5 366 84
XM_017005127.1 XP_016860616.1 4 - 5 1056 84
XM_017005128.1 XP_016860617.1 4 - 5 403 84
XM_017005129.1 XP_016860618.1 3 - 4 571 84
XM_017005130.1 XP_016860619.1 3 - 4 571 84
XM_017005132.1 XP_016860621.1 2 - 3 143 84
XM_017005133.1 XP_016860622.1 4 - 5 573 84
XM_017005134.1 XP_016860623.1 3 - 4 943 84
XM_017005135.1 XP_016860624.1 2 - 3 143 84
GenBank mRNA AK054911.1 - 4 - 5 1133 84
AK055823.1 - - 711 84
AK128720.1 - 2 - 3 452 84
BC048013.1 - 4 - 5 535 84
BC050308.1 - 4 - 5 401 84
X81889.1 - 4 - 5 416 84

View Full Product Details