The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000540.2 | NP_000531.2 | 35 - 36 | 5943 | 71 |
NM_001042723.1 | NP_001036188.1 | 35 - 36 | 5943 | 71 | |
XM_006723317.2 | XP_006723380.1 | 35 - 36 | 5893 | 71 | |
XM_006723319.2 | XP_006723382.1 | 35 - 36 | 5893 | 71 | |
XM_011527205.2 | XP_011525507.1 | 35 - 36 | 5893 | 71 | |
GenBank mRNA | A20359.1 | - | 35 - 36 | 5915 | 71 |
J05200.1 | - | 35 - 36 | 5915 | 71 |