Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RYR1
Entrez Gene ID
6261
Gene Name
ryanodine receptor 1
Gene Aliases
CCO, MHS, MHS1, PPP1R137, RYDR, RYR, RYR-1, SKRR
Chromosome Location
Chr.19: 38433700 - 38587564 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000540.2 NP_000531.2 94 - 95 13865 95
NM_001042723.1 NP_001036188.1 93 - 94 13850 95
XM_006723317.2 XP_006723380.1 93 - 94 13797 95
XM_006723319.2 XP_006723382.1 92 - 93 13782 95
GenBank mRNA A20359.1 - 93 - 94 13822 95
J05200.1 - 93 - 94 13822 95

View Full Product Details