Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRUNE2
Entrez Gene ID
158471
Gene Name
prune homolog 2
Gene Aliases
BMCC1, BNIPXL, C9orf65, KIAA0367
Chromosome Location
Chr.9: 76611376 - 76906220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308049.1 NP_001294978.1 10 - 11 1393 111
NM_001308050.1 NP_001294979.1 9 - 10 1248 111
NM_015225.2 NP_056040.2 17 - 18 9306 111
XM_005251745.1 XP_005251802.1 18 - 19 9451 111
XM_006716982.1 XP_006717045.1 19 - 20 9487 111
XM_006716983.1 XP_006717046.1 19 - 20 9484 111
XM_006716984.2 XP_006717047.1 19 - 20 9487 158
XM_006716985.1 XP_006717048.1 18 - 19 9478 111
XM_006716986.1 XP_006717049.1 18 - 19 9475 111
XM_011518323.1 XP_011516625.1 17 - 18 9442 111
XM_017014345.1 XP_016869834.1 18 - 19 9448 111
XM_017014354.1 XP_016869843.1 10 - 11 1076 111
XM_017014355.1 XP_016869844.1 9 - 10 1040 111
XM_017014358.1 XP_016869847.1 9 - 10 2894 111
GenBank mRNA AB002365.2 - 10 - 11 2378 111
AB050197.1 - 11 - 12 8412 111
AK296269.1 - 10 - 11 1175 111
AK298805.1 - 9 - 10 1030 111
AY439213.1 - 11 - 12 2387 111
BC150641.1 - 11 - 12 8324 111

View Full Product Details