The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003886.2 | NP_003877.2 | 1 - 2 | 155 | 68 |
GenBank mRNA | AK292463.1 | - | 1 - 2 | 149 | 68 |
BC126250.1 | - | 1 - 2 | 154 | 68 | |
BC126252.1 | - | 1 - 2 | 154 | 68 | |
Y15195.1 | - | 1 - 2 | 155 | 68 |