The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323906.1 | NP_001310835.1 | 5 - 6 | 982 | 88 |
NM_001323908.1 | NP_001310837.1 | 5 - 6 | 1014 | 88 | |
NM_001323910.1 | NP_001310839.1 | 6 - 7 | 1030 | 88 | |
NM_001323913.1 | NP_001310842.1 | 4 - 5 | 814 | 88 | |
NM_144618.2 | NP_653219.1 | 5 - 6 | 956 | 88 | |
XM_017000246.1 | XP_016855735.1 | 6 - 7 | 1460 | 88 | |
XM_017000247.1 | XP_016855736.1 | 7 - 8 | 1547 | 88 | |
XM_017000248.1 | XP_016855737.1 | 5 - 6 | 856 | 88 | |
XM_017000249.1 | XP_016855738.1 | 6 - 7 | 916 | 88 | |
XM_017000252.1 | XP_016855741.1 | 5 - 6 | 702 | 88 | |
XM_017000253.1 | XP_016855742.1 | 5 - 6 | 673 | 88 | |
GenBank mRNA | AK092230.1 | - | 5 - 6 | 796 | 88 |
AK301882.1 | - | 6 - 7 | 794 | 88 | |
AK313606.1 | - | 5 - 6 | 769 | 88 | |
BC027033.1 | - | 5 - 6 | 956 | 88 |