The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013939.2 | NP_039227.1 | 1 - 1 | 578 | 107 |
GenBank mRNA | BC069085.1 | - | 1 - 1 | 558 | 107 |
BC069457.1 | - | 1 - 1 | 558 | 107 | |
BC112178.1 | - | 1 - 1 | 590 | 107 | |
BC113647.1 | - | 1 - 1 | 578 | 107 |