Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
Chromosome Location
Chr.19: 17747718 - 17788568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161357.1 NP_001154829.1 14 - 15 1228 78
NM_001161358.1 NP_001154830.1 13 - 14 1138 78
NM_001161359.1 NP_001154831.1 12 - 13 1068 78
NM_015122.2 NP_055937.1 14 - 15 1232 78
XM_005259831.3 XP_005259888.1 13 - 14 1317 78
XM_006722701.3 XP_006722764.1 13 - 14 1236 78
XM_006722702.3 XP_006722765.1 13 - 14 1313 78
XM_011527824.2 XP_011526126.1 14 - 15 1228 78
XM_011527825.2 XP_011526127.1 14 - 15 1217 78
XM_011527826.2 XP_011526128.1 14 - 15 1200 78
XM_011527827.2 XP_011526129.1 12 - 13 1035 78
XM_011527828.2 XP_011526130.1 13 - 14 1131 78
XM_017026519.1 XP_016882008.1 14 - 15 1298 78
XM_017026520.1 XP_016882009.1 14 - 15 1281 78
XM_017026521.1 XP_016882010.1 13 - 14 1232 78
XM_017026522.1 XP_016882011.1 14 - 15 1228 78
XM_017026523.1 XP_016882012.1 14 - 15 1298 78
XM_017026524.1 XP_016882013.1 13 - 14 1313 78
XM_017026525.1 XP_016882014.1 13 - 14 1111 78
XM_017026526.1 XP_016882015.1 14 - 15 1228 78
XM_017026527.1 XP_016882016.1 13 - 14 1236 78
XM_017026528.1 XP_016882017.1 14 - 15 1298 78
GenBank mRNA AB006628.1 - 11 - 12 1002 78
AK291410.1 - 13 - 14 1094 78
AK303623.1 - 10 - 11 930 78
AK309403.1 - 10 - 11 981 78
AK316458.1 - 12 - 13 1039 78
BC028021.1 - 14 - 15 1173 78
BC041130.1 - 14 - 15 1182 78

View Full Product Details