Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
Chromosome Location
Chr.19: 17747718 - 17788568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161357.1 NP_001154829.1 23 - 24 2218 90
NM_001161358.1 NP_001154830.1 22 - 23 2128 90
NM_001161359.1 NP_001154831.1 21 - 22 2058 90
NM_015122.2 NP_055937.1 23 - 24 2222 90
XM_005259831.3 XP_005259888.1 22 - 23 2307 90
XM_006722701.3 XP_006722764.1 22 - 23 2226 90
XM_006722702.3 XP_006722765.1 22 - 23 2303 90
XM_011527824.2 XP_011526126.1 23 - 24 2218 90
XM_011527825.2 XP_011526127.1 23 - 24 2207 90
XM_011527826.2 XP_011526128.1 23 - 24 2190 90
XM_011527827.2 XP_011526129.1 20 - 21 1749 90
XM_011527828.2 XP_011526130.1 22 - 23 2121 90
XM_017026519.1 XP_016882008.1 23 - 24 2288 90
XM_017026520.1 XP_016882009.1 23 - 24 2271 90
XM_017026521.1 XP_016882010.1 22 - 23 2222 90
XM_017026522.1 XP_016882011.1 23 - 24 2101 90
XM_017026523.1 XP_016882012.1 23 - 24 2171 90
XM_017026524.1 XP_016882013.1 22 - 23 2186 90
XM_017026525.1 XP_016882014.1 22 - 23 1984 90
XM_017026526.1 XP_016882015.1 22 - 23 1942 90
XM_017026527.1 XP_016882016.1 21 - 22 1950 90
XM_017026528.1 XP_016882017.1 22 - 23 2012 90
GenBank mRNA AB006628.1 - 20 - 21 1992 90
AK291410.1 - 22 - 23 2084 90
AK303623.1 - 19 - 20 1920 90
AK309403.1 - 19 - 20 1971 90
AK316458.1 - 21 - 22 2029 90
BC028021.1 - 23 - 24 2163 90
BC041130.1 - 23 - 24 2172 90

View Full Product Details