Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
Chromosome Location
Chr.19: 17747718 - 17788568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161357.1 NP_001154829.1 24 - 25 2367 78
NM_001161358.1 NP_001154830.1 23 - 24 2277 78
NM_001161359.1 NP_001154831.1 22 - 23 2207 78
NM_015122.2 NP_055937.1 24 - 25 2371 78
XM_005259831.3 XP_005259888.1 23 - 24 2456 78
XM_006722701.3 XP_006722764.1 23 - 24 2375 78
XM_006722702.3 XP_006722765.1 23 - 24 2452 78
XM_011527824.2 XP_011526126.1 24 - 25 2367 78
XM_011527825.2 XP_011526127.1 24 - 25 2356 78
XM_011527826.2 XP_011526128.1 24 - 25 2339 78
XM_011527827.2 XP_011526129.1 21 - 22 1898 78
XM_011527828.2 XP_011526130.1 23 - 24 2270 78
XM_017026519.1 XP_016882008.1 24 - 25 2437 78
XM_017026520.1 XP_016882009.1 24 - 25 2420 78
XM_017026521.1 XP_016882010.1 23 - 24 2371 78
XM_017026522.1 XP_016882011.1 24 - 25 2250 78
XM_017026523.1 XP_016882012.1 24 - 25 2320 78
XM_017026524.1 XP_016882013.1 23 - 24 2335 78
XM_017026525.1 XP_016882014.1 23 - 24 2133 78
XM_017026526.1 XP_016882015.1 23 - 24 2091 78
XM_017026527.1 XP_016882016.1 22 - 23 2099 78
XM_017026528.1 XP_016882017.1 23 - 24 2161 78
GenBank mRNA AB006628.1 - 21 - 22 2141 78
AK291410.1 - 23 - 24 2233 78
AK303623.1 - 20 - 21 2069 78
AK309403.1 - 20 - 21 2120 78
AK316458.1 - 22 - 23 2178 78
BC028021.1 - 24 - 25 2312 78
BC041130.1 - 24 - 25 2321 78

View Full Product Details