Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
Chromosome Location
Chr.19: 17747718 - 17788568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161357.1 NP_001154829.1 26 - 27 2703 61
NM_001161358.1 NP_001154830.1 25 - 26 2613 61
NM_001161359.1 NP_001154831.1 24 - 25 2543 61
NM_015122.2 NP_055937.1 26 - 27 2707 61
XM_005259831.3 XP_005259888.1 25 - 26 2792 61
XM_006722701.3 XP_006722764.1 25 - 26 2711 61
XM_006722702.3 XP_006722765.1 25 - 26 2788 61
XM_011527824.2 XP_011526126.1 26 - 27 2703 61
XM_011527825.2 XP_011526127.1 26 - 27 2692 61
XM_011527826.2 XP_011526128.1 26 - 27 2675 61
XM_011527827.2 XP_011526129.1 23 - 24 2234 61
XM_011527828.2 XP_011526130.1 25 - 26 2606 61
XM_017026519.1 XP_016882008.1 26 - 27 2773 61
XM_017026520.1 XP_016882009.1 26 - 27 2756 61
XM_017026521.1 XP_016882010.1 25 - 26 2707 61
XM_017026522.1 XP_016882011.1 26 - 27 2586 61
XM_017026523.1 XP_016882012.1 26 - 27 2656 61
XM_017026524.1 XP_016882013.1 25 - 26 2671 61
XM_017026525.1 XP_016882014.1 25 - 26 2469 61
XM_017026526.1 XP_016882015.1 25 - 26 2427 61
XM_017026527.1 XP_016882016.1 24 - 25 2435 61
XM_017026528.1 XP_016882017.1 25 - 26 2497 61
GenBank mRNA AB006628.1 - 23 - 24 2477 61
AK291410.1 - 25 - 26 2569 61
AK303623.1 - 22 - 23 2405 61
AK309403.1 - 22 - 23 2456 61
AK316458.1 - 24 - 25 2514 61
BC028021.1 - 26 - 27 2648 61
BC041130.1 - 26 - 27 2657 61

View Full Product Details