The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024690.2 | NP_078966.2 | 68 - 69 | 42081 | 64 |
XM_017027486.1 | XP_016882975.1 | 62 - 63 | 41161 | 64 | |
XM_017027487.1 | XP_016882976.1 | 61 - 62 | 41095 | 64 | |
XM_017027488.1 | XP_016882977.1 | 62 - 63 | 41161 | 64 | |
XM_017027489.1 | XP_016882978.1 | 59 - 60 | 41007 | 64 | |
XM_017027490.1 | XP_016882979.1 | 57 - 58 | 40696 | 64 | |
XM_017027491.1 | XP_016882980.1 | 57 - 58 | 40696 | 64 | |
XM_017027492.1 | XP_016882981.1 | 57 - 58 | 40693 | 64 | |
XM_017027493.1 | XP_016882982.1 | 57 - 58 | 40693 | 64 | |
XM_017027494.1 | XP_016882983.1 | 57 - 58 | 40693 | 64 | |
XM_017027495.1 | XP_016882984.1 | 57 - 58 | 40693 | 64 | |
XM_017027496.1 | XP_016882985.1 | 42 - 43 | 39292 | 64 | |
XM_017027497.1 | XP_016882986.1 | 37 - 38 | 38818 | 64 | |
XM_017027499.1 | XP_016882988.1 | 62 - 63 | 31948 | 64 | |
XM_017027500.1 | XP_016882989.1 | 61 - 62 | 31867 | 64 | |
XM_017027501.1 | XP_016882990.1 | 41 - 42 | 3875 | 64 | |
GenBank mRNA | AF361486.3 | - | - | 19341 | 64 |
AF414442.2 | - | - | 64 | ||
AK024365.1 | - | 20 - 21 | 64 | ||
AK128681.1 | - | 18 - 19 | 2460 | 64 | |
AK128856.1 | - | 25 - 26 | 2501 | 64 | |
AK128857.1 | - | - | 64 |