Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MUC16
Entrez Gene ID
94025
Gene Name
mucin 16, cell surface associated
Gene Aliases
CA125
Chromosome Location
Chr.19: 8848840 - 9010390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024690.2 NP_078966.2 73 - 74 42532 79
XM_017027486.1 XP_016882975.1 67 - 68 41612 79
XM_017027487.1 XP_016882976.1 66 - 67 41546 79
XM_017027488.1 XP_016882977.1 65 - 66 41525 79
XM_017027489.1 XP_016882978.1 64 - 65 41458 79
XM_017027490.1 XP_016882979.1 62 - 63 41147 79
XM_017027491.1 XP_016882980.1 62 - 63 41147 79
XM_017027492.1 XP_016882981.1 62 - 63 41144 79
XM_017027493.1 XP_016882982.1 62 - 63 41144 79
XM_017027494.1 XP_016882983.1 62 - 63 41144 79
XM_017027495.1 XP_016882984.1 62 - 63 41144 79
XM_017027496.1 XP_016882985.1 47 - 48 39743 79
XM_017027497.1 XP_016882986.1 42 - 43 39269 79
XM_017027499.1 XP_016882988.1 67 - 68 32399 79
XM_017027500.1 XP_016882989.1 66 - 67 32318 79
XM_017027501.1 XP_016882990.1 46 - 47 4326 79
GenBank mRNA AF361486.3 - - 19792 79
AF414442.2 - - 65467 79
AK024365.1 - 25 - 26 2275 79
AK128856.1 - 29 - 30 2901 79
AK128857.1 - - 4162 79

View Full Product Details