The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291411.1 | NP_001278340.1 | 2 - 3 | 300 | 158 |
NM_032195.2 | NP_115571.2 | 2 - 3 | 300 | 158 | |
NM_138927.2 | NP_620305.2 | 2 - 3 | 300 | 158 | |
NR_103797.1 | 2 - 3 | 300 | 158 | ||
GenBank mRNA | AB028942.2 | - | 2 - 3 | 269 | 158 |
AF161428.1 | - | 2 - 3 | 275 | 158 | |
AF161430.1 | - | 2 - 3 | 271 | 158 | |
AF380179.1 | - | 2 - 3 | 274 | 158 | |
AF380180.1 | - | 2 - 3 | 274 | 158 | |
AF380181.1 | - | 2 - 3 | 274 | 158 | |
AF380183.1 | - | 2 - 3 | 274 | 158 | |
AF380184.1 | - | 2 - 3 | 274 | 158 | |
AF435977.1 | - | 2 - 3 | 260 | 158 | |
AK024752.1 | - | 2 - 3 | 279 | 158 | |
AK307612.1 | - | 2 - 3 | 300 | 158 | |
AY026895.1 | - | 2 - 3 | 294 | 158 | |
BC005337.1 | - | 2 - 3 | 276 | 158 | |
BC029455.1 | - | 2 - 3 | 720 | 158 | |
BC054029.1 | - | 2 - 3 | 254 | 158 | |
BC066303.1 | - | 2 - 3 | 268 | 158 | |
BC104656.1 | - | 2 - 3 | 365 | 158 |