The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_103797.1 | 9 - 10 | 7017 | 80 | |
GenBank mRNA | AF380181.1 | - | 9 - 10 | 6991 | 80 |
AF380182.1 | - | 1 - 2 | 259 | 80 |