The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144072.1 | NP_001137544.1 | 3 - 4 | 759 | 148 |
NR_026644.1 | 3 - 4 | 903 | 148 | ||
XM_006719948.3 | XP_006720011.1 | 4 - 5 | 469 | 148 | |
XM_011521084.2 | XP_011519386.1 | 3 - 4 | 324 | 148 | |
GenBank mRNA | AK075419.1 | - | 3 - 4 | 330 | 148 |
AK129861.1 | - | 3 - 4 | 358 | 148 | |
BC121138.1 | - | 3 - 4 | 502 | 148 | |
BC121139.1 | - | 3 - 4 | 502 | 148 |