Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MFF
- Entrez Gene ID
-
56947
- Gene Name
- mitochondrial fission factor
- Gene Aliases
-
C2orf33, GL004
- Chromosome Location
-
Chr.2: 227325151 - 227357836 on Build GRCh38
View Full Product Details