Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MDM2
- Entrez Gene ID
-
4193
- Gene Name
- MDM2 proto-oncogene
- Gene Aliases
-
ACTFS, HDMX, hdm2
- Chromosome Location
-
Chr.12: 68808149 - 68845544 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U33200.1 |
- |
1 - 2 |
79 |
115 |
View Full Product Details