Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MDM2
Entrez Gene ID
4193
Gene Name
MDM2 proto-oncogene
Gene Aliases
ACTFS, HDMX, hdm2
Chromosome Location
Chr.12: 68808149 - 68845544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF201370.1 - 1 - 2 65 70
U33199.1 - 1 - 2 80 70

View Full Product Details