The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012094.4 | NP_036226.1 | 2 - 3 | 432 | 80 |
GenBank mRNA | AF110731.1 | - | 2 - 3 | 340 | 80 |
AF112212.1 | - | 2 - 3 | 331 | 80 | |
AF124993.1 | - | 2 - 3 | 373 | 80 | |
AF197952.1 | - | 2 - 3 | 413 | 80 | |
AF231705.1 | - | 2 - 3 | 323 | 80 | |
AF242525.1 | - | 2 - 3 | 339 | 80 | |
AJ249483.1 | - | 1 - 2 | 148 | 80 | |
BC110983.1 | - | 2 - 3 | 432 | 80 | |
BC113723.1 | - | 2 - 3 | 338 | 80 | |
BC113725.1 | - | 2 - 3 | 338 | 80 | |
BU598032.1 | - | 2 - 3 | 426 | 80 | |
CR457203.1 | - | 2 - 3 | 304 | 80 | |
FJ224298.1 | - | 1 - 2 | 148 | 80 |