The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014510.2 | NP_055325.2 | 10 - 11 | 13939 | 150 |
NM_033026.5 | NP_149015.2 | 10 - 11 | 13939 | 150 | |
XM_017012006.1 | XP_016867495.1 | 8 - 9 | 8119 | 150 | |
XM_017012007.1 | XP_016867496.1 | 8 - 9 | 8103 | 150 | |
GenBank mRNA | AB011131.1 | - | 4 - 5 | 2483 | 150 |
BC001304.1 | - | 2 - 3 | 180 | 150 | |
BC108289.1 | - | 1 - 2 | 141 | 150 |