Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHD9
Entrez Gene ID
80205
Gene Name
chromodomain helicase DNA binding protein 9
Gene Aliases
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location
Chr.16: 53054991 - 53327502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308319.1 NP_001295248.1 1 - 2 1617 107
NM_025134.4 NP_079410.4 2 - 3 1662 107
XM_005256168.3 XP_005256225.1 2 - 3 1704 107
XM_005256169.2 XP_005256226.1 2 - 3 1726 107
XM_005256172.3 XP_005256229.1 2 - 3 1643 107
XM_006721281.2 XP_006721344.1 2 - 3 1643 107
XM_006721283.3 XP_006721346.1 2 - 3 1864 107
XM_011523347.2 XP_011521649.1 3 - 4 1776 107
XM_017023718.1 XP_016879207.1 3 - 4 1899 107
XM_017023719.1 XP_016879208.1 4 - 5 1850 107
XM_017023720.1 XP_016879209.1 3 - 4 1744 107
XM_017023721.1 XP_016879210.1 3 - 4 1900 107
XM_017023722.1 XP_016879211.1 3 - 4 1900 107
XM_017023723.1 XP_016879212.1 3 - 4 1900 107
XM_017023728.1 XP_016879217.1 2 - 3 1702 107
GenBank mRNA AB002306.3 - 1 - 2 1085 107
AK092208.1 - 2 - 3 1726 107
AY243500.1 - 1 - 2 1540 107
AY647157.1 - 2 - 3 1662 107
BC033770.1 - 2 - 3 1672 107
BC140815.1 - 1 - 2 1494 107
BC144622.1 - 1 - 2 1494 107
DQ333316.1 - 2 - 3 1756 107

View Full Product Details