Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHROOM3
Entrez Gene ID
57619
Gene Name
shroom family member 3
Gene Aliases
APXL3, MSTP013, SHRM, ShrmL
Chromosome Location
Chr.4: 76435100 - 76783253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020859.3 NP_065910.3 4 - 5 1537 64
XM_005263163.4 XP_005263220.1 3 - 4 5991 64
XM_005263164.4 XP_005263221.1 3 - 4 549 64
XM_005263165.4 XP_005263222.1 3 - 4 1975 64
XM_006714282.3 XP_006714345.1 4 - 5 692 64
XM_011532157.2 XP_011530459.2 3 - 4 1379 64
XM_011532158.2 XP_011530460.1 5 - 6 1338 64
XM_011532159.1 XP_011530461.1 4 - 5 1527 64
XM_011532160.1 XP_011530462.1 2 - 3 539 64
XM_011532161.2 XP_011530463.1 3 - 4 717 64
XM_017008488.1 XP_016863977.1 4 - 5 695 64
XM_017008489.1 XP_016863978.1 4 - 5 596 64
GenBank mRNA AB055660.1 - 4 - 5 1538 64
AK127929.1 - 2 - 3 539 64
AK304103.1 - 3 - 4 333 64

View Full Product Details