The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005956.3 | NP_005947.3 | 4 - 5 | 626 | 112 |
GenBank mRNA | AK302747.1 | - | 4 - 5 | 636 | 112 |
AK309266.1 | - | 3 - 4 | 239 | 112 | |
AK312361.1 | - | 4 - 5 | 331 | 112 | |
BC009806.2 | - | 4 - 5 | 292 | 112 | |
BC050420.1 | - | 4 - 5 | 317 | 112 | |
BQ959308.1 | - | 4 - 5 | 438 | 112 | |
J04031.1 | - | 4 - 5 | 292 | 112 |