Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS12
Entrez Gene ID
57117
Gene Name
integrator complex subunit 12
Gene Aliases
INT12, PHF22, SBBI22
Chromosome Location
Chr.4: 105682627 - 105708725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142471.1 NP_001135943.1 2 - 3 252 89
NM_020395.3 NP_065128.2 3 - 4 414 89
XM_005263148.4 XP_005263205.1 2 - 3 907 89
XM_011532143.2 XP_011530445.1 3 - 4 1063 89
XM_011532145.2 XP_011530447.1 4 - 5 550 89
GenBank mRNA AF242524.1 - 2 - 3 246 89
AK314939.1 - 2 - 3 252 89
BC014442.1 - 3 - 4 362 89

View Full Product Details