Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS12
Entrez Gene ID
57117
Gene Name
integrator complex subunit 12
Gene Aliases
INT12, PHF22, SBBI22
Chromosome Location
Chr.4: 105682627 - 105708725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142471.1 NP_001135943.1 4 - 5 594 74
NM_020395.3 NP_065128.2 5 - 6 756 74
XM_005263148.4 XP_005263205.1 4 - 5 1249 74
XM_011532143.2 XP_011530445.1 5 - 6 1405 74
XM_011532145.2 XP_011530447.1 6 - 7 892 74
GenBank mRNA AF242524.1 - 4 - 5 588 74
AK314939.1 - 4 - 5 594 74
BC014442.1 - 5 - 6 704 74

View Full Product Details