The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160130.1 | NP_001153602.1 | 7 - 8 | 1474 | 61 |
NM_001160132.1 | NP_001153604.1 | 7 - 8 | 1474 | 61 | |
NM_001160133.1 | NP_001153605.1 | 7 - 8 | 1474 | 61 | |
NM_001160134.1 | NP_001153606.1 | 7 - 8 | 1474 | 61 | |
NM_019842.3 | NP_062816.2 | 7 - 8 | 1474 | 61 | |
XM_017011058.1 | XP_016866547.1 | 7 - 8 | 1474 | 61 | |
GenBank mRNA | AF202977.1 | - | 7 - 8 | 1022 | 61 |
AF249278.1 | - | 7 - 8 | 1236 | 61 | |
AF263835.1 | - | 7 - 8 | 914 | 61 | |
AK299550.1 | - | 7 - 8 | 1074 | 61 | |
BC050689.1 | - | 7 - 8 | 1211 | 61 | |
BC117359.1 | - | 7 - 8 | 1145 | 61 | |
BC143554.1 | - | 7 - 8 | 1145 | 61 | |
BC143555.1 | - | 6 - 7 | 1018 | 61 |