The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019842.3 | NP_062816.2 | 9 - 10 | 1596 | 71 |
GenBank mRNA | AF202977.1 | - | 9 - 10 | 1144 | 71 |
AF249278.1 | - | 9 - 10 | 1358 | 71 | |
AF263835.1 | - | 9 - 10 | 1036 | 71 |