Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEF1
Entrez Gene ID
553115
Gene Name
penta-EF-hand domain containing 1
Gene Aliases
ABP32, PEF1A
Chromosome Location
Chr.1: 31629862 - 31645237 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012392.3 NP_036524.1 2 - 3 696 85
NR_033688.1 2 - 3 537 85
XM_011541745.1 XP_011540047.1 2 - 3 323 85
XM_011541746.2 XP_011540048.1 2 - 3 790 85
XM_011541747.1 XP_011540049.1 2 - 3 665 85
XM_017001680.1 XP_016857169.1 2 - 3 654 85
GenBank mRNA AB018357.1 - 2 - 3 335 85
AB026628.1 - 2 - 3 335 85
AK001420.1 - 2 - 3 348 85
AY359011.1 - 2 - 3 339 85
BC002773.2 - 2 - 3 325 85
BC012561.1 - 2 - 3 341 85
BI826532.1 - 2 - 3 509 85
CR542139.1 - 2 - 3 323 85

View Full Product Details