Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D5
Entrez Gene ID
400745
Gene Name
SH2 domain containing 5
Gene Aliases
-
Chromosome Location
Chr.1: 20719731 - 20732640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103160.1 NP_001096630.1 8 - 9 1443 98
NM_001103161.1 NP_001096631.1 9 - 10 1572 98
XM_011541459.1 XP_011539761.1 9 - 10 2037 98
XM_011541460.1 XP_011539762.1 10 - 11 1877 98
XM_011541461.2 XP_011539763.1 9 - 10 1150 98
XM_011541462.1 XP_011539764.1 9 - 10 2034 98
GenBank mRNA AK123841.1 - 5 - 6 801 98
AK124869.1 - 8 - 9 1443 98
AK296431.1 - 9 - 10 1141 98

View Full Product Details