The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003568.2 | NP_003559.2 | 2 - 3 | 455 | 83 |
XM_011510058.2 | XP_011508360.1 | 2 - 3 | 174 | 83 | |
GenBank mRNA | AF230929.1 | - | 2 - 3 | 211 | 83 |
AJ009985.1 | - | 2 - 3 | 399 | 83 | |
AK314926.1 | - | 2 - 3 | 174 | 83 | |
BC005830.2 | - | 1 - 2 | 111 | 83 | |
BJ993162.1 | - | 2 - 3 | 455 | 83 |