Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRAM2
Entrez Gene ID
128338
Gene Name
DNA damage regulated autophagy modulator 2
Gene Aliases
CORD21, PRO180, TMEM77, WWFQ154
Chromosome Location
Chr.1: 111117332 - 111140216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178454.4 NP_848549.3 6 - 7 774 66
XM_005270469.2 XP_005270526.1 7 - 8 832 66
XM_005270470.1 XP_005270527.1 7 - 8 955 66
XM_006710361.1 XP_006710424.1 7 - 8 792 66
XM_006710362.1 XP_006710425.1 7 - 8 765 66
XM_011540707.1 XP_011539009.1 7 - 8 773 66
XM_011540708.1 XP_011539010.1 7 - 8 853 66
XM_017000320.1 XP_016855809.1 6 - 7 716 66
XM_017000321.1 XP_016855810.1 6 - 7 575 66
GenBank mRNA AK075350.1 - 2 - 3 220 66
AL833098.1 - 6 - 7 1142 66
AY336747.1 - 6 - 7 652 66
AY358492.1 - 6 - 7 637 66
BC047025.1 - 5 - 6 589 66
BC091509.1 - 7 - 8 866 66
EF710624.1 - 4 - 5 517 66

View Full Product Details