The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173814.5 | NP_776175.2 | 10 - 11 | 2034 | 69 |
XM_011521459.1 | XP_011519761.1 | 10 - 11 | 2564 | 69 | |
XM_011521460.1 | XP_011519762.1 | 10 - 11 | 2564 | 69 | |
XM_017022081.1 | XP_016877570.1 | 10 - 11 | 2121 | 69 | |
XM_017022082.1 | XP_016877571.1 | 10 - 11 | 2564 | 69 | |
GenBank mRNA | AK098622.1 | - | 1 - 2 | 42 | 69 |
AY630258.1 | - | 10 - 11 | 1894 | 69 |