The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173814.5 | NP_776175.2 | 18 - 19 | 3277 | 167 |
XM_017022081.1 | XP_016877570.1 | 18 - 19 | 3364 | 167 | |
GenBank mRNA | AK098622.1 | - | 9 - 10 | 1285 | 167 |
AY630258.1 | - | 18 - 19 | 3137 | 167 |